missing translation for 'onlineSavingsMsg'
Learn More

o-Cresol, 99%, AcroSeal™

Product Code. 10503648 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10503648 100 mL AcroSeal™ Glass Bottle 100mL
10278127 800 mL AcroSeal™ Glass Bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10503648 Supplier Thermo Scientific Acros Supplier No. 405731000
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 95-48-7 | C7H8O | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-48-7 , 108-95-2
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002226
InChI Key QWVGKYWNOKOFNN-UHFFFAOYSA-N
Synonym o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene
PubChem CID 335
ChEBI CHEBI:28054
IUPAC Name 2-methylphenol
SMILES CC1=CC=CC=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 30.0°C to 32.0°C
Color Amber to Colorless
Density 1.0400g/mL
Boiling Point 191.0°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3C6H4OH
Quantity 100 mL
Beilstein 06,349
Merck Index 15,2564
Specific Gravity 1.04
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether and chloroform
Viscosity 4.75 mPa.s (35°C)
Formula Weight 108.14
Percent Purity 99%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material o-Cresol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • o-Cresol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: o-Cresol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.