missing translation for 'onlineSavingsMsg'
Learn More

o-Cresol, ≥99, Reagent Grade, Honeywell™

Product Code. 15607780 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15607780 100 g Glass bottle 100g
15617780 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15607780 Supplier Honeywell Research Chemicals Supplier No. C85700100G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 95-48-7
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002226
InChI Key QWVGKYWNOKOFNN-UHFFFAOYSA-N
Synonym o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene
PubChem CID 335
ChEBI CHEBI:28054
IUPAC Name 2-methylphenol
SMILES CC1=CC=CC=C1O

Specifications

Melting Point 30.99°C (1013hPa)
Color Colorless
Density 1.05g/cu.cm at 20°C
Boiling Point 191°C (1013 hPa)
Flash Point 81°C (177.8°F)
Ignition Point 1110°F
Odor Characteristic
Packaging Glass bottle
Linear Formula CH3C6H4OH
Quantity 100 g
UN Number UN3455
Beilstein 506917
Solubility Information 26g/L at 25°C Solubility in water
Vapor Density 3.72 (vs air)
Formula Weight 108.14g/mol
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material o-Cresol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.