missing translation for 'onlineSavingsMsg'
Learn More

o-Cresol, TRC

Product Code. 18013250
Change view
Click to view available options
Quantity:
50 g
500 g
Unit Size:
500g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18013250 50 g 50g
18012950 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18013250 Supplier Toronto Research Chemicals Supplier No. TRCC78191050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-48-7
Molecular Formula C7 H8 O
Molecular Weight (g/mol) 108.14
Synonym Phenol, 2-methyl- (9CI, ACI), 2-Methylphenol (ACI), o-Cresol (8CI), 1-Hydroxy-2-methylbenzene, 1-Methyl-2-hydroxybenzene, 2-Cresol, 2-Hydroxytoluene, NSC 23076, NSC 36809, o-Cresylic acid, o-Hydroxytoluene, o-Methylphenol, o-Methylphenylol, o-Oxytoluene, o-Toluol
IUPAC Name 2-methylphenol
SMILES Cc1ccccc1O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C7H8O/c1-6-4-2-3-5-7(6)8/h2-5,8H,1H3
Formula Weight 108.0575
Percent Purity >95%
Chemical Name or Material 2-Methylphenol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.