missing translation for 'onlineSavingsMsg'
Learn More

o-Cresolglycolic Acid, TRC

Product Code. 30137430
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137430 500 mg 500mg
30136138 50 mg 50mg
30009745 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137430 Supplier Toronto Research Chemicals Supplier No. TRCB399870500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-49-5
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.17
Synonym o-Cresoxyacetic Acid, o-Cresyloxyacetic Acid, o-Tolyloxyacetic Acid, 2-(2-Methylphenoxy)acetic Acid, (2-Methylphenoxy)acetic Acid, (o-Methylphenoxy)acetic Acid, 2-(o-Tolyloxy)acetic Acid, 2-Methylphenyloxyacetic Acid, 2-[(2-Methylphenyl)oxy]acetic Acid, NSC 5293;
IUPAC Name 2-(2-methylphenoxy)acetic acid
SMILES Cc1ccccc1OCC(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H10O3/c1-7-4-2-3-5-8(7)12-6-9(10)11/h2-5H,6H2,1H3,(H,10,11)
Formula Weight 166.063
Chemical Name or Material o-Cresolglycolic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.