missing translation for 'onlineSavingsMsg'
Learn More

O,O-Dimethyl Phosphorothionate Ammonium Salt, TRC

Product Code. 30134629
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134629 10 g 10g
30010758 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134629 Supplier Toronto Research Chemicals Supplier No. TRCD47762010G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40633-14-5
Molecular Formula C2 H6 O3 P S . H4 N
Molecular Weight (g/mol) 159.14
Synonym Phosphorothioic acid, O,O-dimethyl ester, ammonium salt (1:1), Ammonium methyl phosphorothioate ((NH4O)(MeO)2PS) (7CI), Phosphorothioic acid, O,O-dimethyl ester, ammonium salt (9CI), Ammonium O,O-dimethyl phosphorothioate, Ammonium O,O-dimethyl thiophosphate, Ammonium dimethyl phosphorothioate
IUPAC Name azanium;[methoxy(sulfido)phosphoryl]oxymethane
SMILES [NH4+].COP(=O)([S-])OC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C2H7O3PS.H3N/c1-4-6(3,7)5-2;/h1-2H3,(H,3,7);1H3
Formula Weight 159.0119
Chemical Name or Material O,O-Dimethyl Phosphorothionate Ammonium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.