missing translation for 'onlineSavingsMsg'
Learn More

O,O',O' '-Triethyl Phosphorothioate, TRC

Product Code. 30007044
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007044 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30007044 Supplier Toronto Research Chemicals Supplier No. TRCT7770101G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 126-68-1
Molecular Formula C6 H15 O3 P S
Quantity 1 g
Synonym Phosphorothioic acid, O,O,O-triethyl ester, Ethyl phosphorothioate, (EtO)3PS (6CI), Ethyl thiophosphate, (EtO)3PS (5CI), NSC 2650, O,O,O-TEPT, O,O,O-Triethyl phosphorothioate, O,O,O-Triethyl thiophosphate, O,O',O' '-Triethyl phosphorothioate, Thiophosphoric acid tri-O-ethyl ester, Triethoxyphosphine sulfide, Triethyl phosphorothioate, Triethyl phosphorothionate, Triethyl thiophosphate
InChI Formula InChI=1S/C6H15O3PS/c1-4-7-10(11,8-5-2)9-6-3/h4-6H2,1-3H3
SMILES CCOP(=S)(OCC)OCC
IUPAC Name triethoxy(sulfanylidene)-λ^{5}-phosphane
Molecular Weight (g/mol) 198.22
Formula Weight 198.048
Chemical Name or Material O,O,O-Triethylphosphorothioate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.