missing translation for 'onlineSavingsMsg'
Learn More

o-Phenanthroline EU Pharmacopoeia 0.1% w/v, Fisher Chemical™

Product Code. 12993274 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12993274 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12993274 Supplier Fisher Chemical Supplier No. RP 337 1000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Packaging Bottle
CAS 66-71-7
Analyte or Component Names o-Phenanthroline
Molecular Formula C12H8N2
Concentration 0.1% w/v
InChI Key DGEZNRSVGBDHLK-UHFFFAOYSA-N
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1
Element o-Phenanthroline
IUPAC Name 1,10-phenanthroline
Solvent or Matrix Water
Molecular Weight (g/mol) 180.21
PubChem CID 1318
Quantity 1000 mL
ChEBI CHEBI:44975
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.