missing translation for 'onlineSavingsMsg'
Learn More

O-Phenylhydroxylamine hydrochloride, 97%

Product Code. 11945671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11945671 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11945671 Supplier Thermo Scientific Acros Supplier No. 438810010
 Restricted Item
Estimated Shipment: 17-06-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6092-80-4 | C6H7NO·ClH | 145.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6092-80-4
CAS Min % 96.0
CAS Max % 100.0
Melting Point 121.0°C to 122.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C6H7NO·ClH
Quantity 1 g
Synonym o-phenylhydroxylamine hydrochloride, o-phenylhydroxylamine hcl, phenoxyamine hydrochloride, hydroxylamine, o-phenyl-, hydrochloride, acmc-20ak08, n-phenoxyamine hydrochloride, o-phenylhydroxylamne hydrochlorde, o-phenyl-hydroxylamine hydrochloride, o-phenylhydroxylamine hydrochloride at
InChI Key DBTXKJJSFWZJNS-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)ON.Cl
IUPAC Name O-phenylhydroxylamine;hydrochloride
Molecular Weight (g/mol) 145.59
PubChem CID 6365179
Formula Weight 145.59
Percent Purity 97%
Physical Form Powder
Chemical Name or Material O-Phenylhydroxylamine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • O-Phenylhydroxylamine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.