missing translation for 'onlineSavingsMsg'
Learn More

o-Phthalaldehyde, TRC

Product Code. 30004576
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004576 5 g 5g
30009684 1 g 1g
30007636 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004576 Supplier Toronto Research Chemicals Supplier No. TRCP3842005G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 643-79-8
Molecular Formula C8 H6 O2
Molecular Weight (g/mol) 134.13
Synonym 1,2-Benzenedicarboxaldehyde, Phthalaldehyde, 1,2-Diformylbenzene, 1,2-Formylbenzene, 1,2-Phthalaldehyde, 2-Formylbenzaldehyde, 2-Phthalaldehyde, Benzene-1,2-dicarbaldehyde, CIDEX OPA, NSC 13394, OP 100S, OP 100SF, Phthalic Aldehyde, Phthalic Dialdehyde, Phthalyldicarboxaldehyde, Phtharal, o-Benzenedicarbaldehyde, o-Diformylbenzene, o-Phthaldialdehyde, o-Phthalic Dialdehyde
IUPAC Name phthalaldehyde
SMILES O=Cc1ccccc1C=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H6O2/c9-5-7-3-1-2-4-8(7)6-10/h1-6H
Formula Weight 134.0368
Percent Purity >95%
Chemical Name or Material o-Phthalaldehyde
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.