missing translation for 'onlineSavingsMsg'
Learn More

o-Terphenyl 10000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30314477
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314477 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314477 Supplier Dr. Ehrenstorfer Supplier No. DREYS09010012DI
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material o-Terphenyl
CAS 84-15-1
Analyte or Component Names o-Terphenyl
Shipping Condition Room Temperature
Molecular Formula C18 H14
Concentration 10000 μg/mL
Certifications/Compliance ISO 17034
Synonym 1,1':2',1''-Terphenyl, o-Terphenyl (8CI), 1,1'-Biphenyl, 2-phenyl-, 1,2-Diphenylbenzene, NSC 6809
InChI Formula InChI=1S/C18H14/c1-3-9-15(10-4-1)17-13-7-8-14-18(17)16-11-5-2-6-12-16/h1-14H
SMILES c1ccc(cc1)c2ccccc2c3ccccc3
IUPAC Name 1,2-diphenylbenzene
Solvent or Matrix Dichloromethane
Industry Type Food Testing
Molecular Weight (g/mol) 230.3
Quantity 5 x 1 mL
Formula Weight 230.1096 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.