missing translation for 'onlineSavingsMsg'
Learn More

o-Thiocresol, 98%

Product Code. 11498700
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498700 100 g 100g
11470627 5 g 5g
11480627 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498700 Supplier Thermo Scientific Alfa Aesar Supplier No. B22460.22
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 137-06-4 | C7H8S | 124.20 g/mol

o-Thiocresol is used as a thiol for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Thiocresol is used as a thiol for proteomics research.

Solubility
Soluble in ether and ethanol. Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-06-4
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004838
InChI Key LXUNZSDDXMPKLP-UHFFFAOYSA-N
Synonym o-toluenethiol, o-thiocresol, 2-methylthiophenol, 2-thiocresol, 2-toluenethiol, o-tolyl mercaptan, o-mercaptotoluene, o-methylthiophenol, o-methylbenzenethiol, benzenethiol, 2-methyl
PubChem CID 8712
IUPAC Name 2-methylbenzenethiol
SMILES CC1=CC=CC=C1S

Specifications

Melting Point 10°C to 12°C
Density 1.054
Boiling Point 195°C
Flash Point 63°C (145°F)
Refractive Index 1.58
Quantity 100 g
UN Number UN2810
Beilstein 774068
Sensitivity Air Sensitive
Merck Index 14,9325
Solubility Information Soluble in ether and ethanol. Insoluble in water.
Formula Weight 124.21
Percent Purity 98%
Chemical Name or Material o-Thiocresol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.