missing translation for 'onlineSavingsMsg'
Learn More

o-Tolidine dihydrochloride, 99%

Product Code. 10581524 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10581524 25 g Glass Bottle 25g
10299600 100 g Glass Bottle 100g
10015660 500 g Glass Bottle 500g
11935271 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10581524 Supplier Thermo Scientific Acros Supplier No. 421130250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 612-82-8 | C14H16N2·2HCl | 285.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-82-8
Molecular Formula C14H16N2·2HCl
Molecular Weight (g/mol) 285.2
InChI Key LUKPNZHXJRJBAN-UHFFFAOYSA-N
Synonym 3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl
PubChem CID 108938
IUPAC Name 4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl

Specifications

Melting Point 130.0°C
Color Beige to Beige-Gray
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (on dry substance) (Argentometry)
Packaging Glass Bottle
Quantity 25 g
Merck Index 15,9675
Formula Weight 285.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material o-Tolidine dihydrochloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • o-Tolidine dihydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
Hazard Statement
  • H302-Harmful if swallowed.
  • H350-May cause cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Sicherheitshinweis
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Ergänzende Informationen
  • MIXTURE LIST-Contain: 3,3-Dimethylbenzidine dihydrochloride

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.