missing translation for 'onlineSavingsMsg'
Learn More

o-Tolidine dihydrochloride, ACS reagent

Product Code. 15254248 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15254248 25 g 25g
15264248 100 g 100g
15274248 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15254248 Supplier Thermo Scientific Acros Supplier No. 449660250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 612-82-8 | C14H16N2·2HCl | 285.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-82-8
Molecular Formula C14H16N2·2HCl
Molecular Weight (g/mol) 285.2
InChI Key LUKPNZHXJRJBAN-UHFFFAOYSA-N
Synonym 3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl
PubChem CID 108938
IUPAC Name 4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl

Specifications

Melting Point 130.0°C
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Quantity 25 g
Merck Index 15,9675
Solubility Information Passes test
Formula Weight 285.2
Grade ACS Reagent
Chemical Name or Material o-Tolidine dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.