missing translation for 'onlineSavingsMsg'
Learn More

o-Tolualdehyde, 98%, stabilized

Product Code. 10489743 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Packaging unitSize
10489743 25 g Glass bottle 25g
10716001 100 g Glass bottle 100g
10387800 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10489743 Proveedor Thermo Scientific Acros N.º de proveedor 152430250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 529-20-4 | C8H8O | 120.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 529-20-4 , 123-31-9
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00003338
InChI Key BTFQKIATRPGRBS-UHFFFAOYSA-N
Synonym o-tolualdehyde, 2-tolualdehyde, 2-formyltoluene, o-toluylaldehyde, o-toluic aldehyde, o-tolylaldehyde, o-methylbenzaldehyde, benzaldehyde, 2-methyl, 2-methyl-benzaldehyde, o-methylbenazldehyde
PubChem CID 10722
ChEBI CHEBI:27434
IUPAC Name 2-methylbenzaldehyde
SMILES CC1=CC=CC=C1C=O

Especificaciones

Melting Point -35°C
Color Colorless to Yellow
Density 1.0390g/mL
Boiling Point 199°C to 200°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CHO
Refractive Index 1.5460 to 1.548
Quantity 25 g
Beilstein 07, 295
Merck Index 15, 9686
Specific Gravity 1.039
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol and ether
Stabilizer 0.1% hydroquinone
Formula Weight 120.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material o-Tolualdehyde, 98%
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • o-Tolualdehyde, stabilized
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.