missing translation for 'onlineSavingsMsg'
Learn More

o-Toluenesulfonamide, 98%

Product Code. 11478917
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478917 50 g 50g
11468917 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478917 Supplier Thermo Scientific Alfa Aesar Supplier No. B20544.18
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 88-19-7 | C7H9NO2S | 171.214 g/mol

o-Toluene sulfonamide was mainly produced as a chemical intermediate for the production of saccharin. A mixture of o-Toluene sulfonamide with the p-isomer is used as a plasticizer for hot -melt adhesives, a chemical intermediate for fluorescent pigments and a chemical intermediate for plasticizer resins. (S080800). Studies suggest that o-Toluene sulfonamide has potential mutagenic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Toluene sulfonamide was mainly produced as a chemical intermediate for the production of saccharin. A mixture of o-Toluene sulfonamide with the p-isomer is used as a plasticizer for hot -melt adhesives, a chemical intermediate for fluorescent pigments and a chemical intermediate for plasticizer resins. (S080800). Studies suggest that o-Toluene sulfonamide has potential mutagenic activity.

Solubility
Soluble in water (1.6 g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-19-7
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.214
MDL Number MFCD00007934
InChI Key YCMLQMDWSXFTIF-UHFFFAOYSA-N
Synonym o-toluenesulfonamide, 2-methylbenzene-1-sulfonamide, benzenesulfonamide, 2-methyl, 2-toluenesulfonamide, o-methylbenzenesulfonamide, toluene-2-sulfonamide, ketjenflex 9, toluene-2-sulphonamide, ortho-toluenesulfonamide, 2-tolylsulfonamide
PubChem CID 6924
IUPAC Name 2-methylbenzenesulfonamide
SMILES CC1=CC=CC=C1S(=O)(=O)N

Specifications

Melting Point 156°C
Boiling Point 210°C (1 mmHg)
Flash Point 178°C (352°F)
Quantity 50 g
Beilstein 1102362
Solubility Information Soluble in water (1.6g/L at 25°C).
Formula Weight 171.22
Percent Purity 99%
Chemical Name or Material o-Toluenesulfonamide
compliance-icons
Product Identifier
  • o-Toluenesulfonamide
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: o-Toluenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.