missing translation for 'onlineSavingsMsg'
Learn More

o-Toluic hydrazide, 98+%

Product Code. 11330164
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11330164 5 g 5g
11340164 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11330164 Supplier Thermo Scientific Alfa Aesar Supplier No. L00668.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7658-80-2 | C8H10N2O | 150.181 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7658-80-2
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00014756
InChI Key KFXLXEQCRFGDRU-UHFFFAOYSA-N
Synonym o-toluic hydrazide, o-toluic acid hydrazide, o-toluohydrazide, 2-toluic hydrazide, 2-methyl-benzoic acid hydrazide, benzoylhydrazine, o-methyl, benzoic acid, 2-methyl-, hydrazide, 2-methylbenzenecarbohydrazide, methylbenzohydrazide, 2-methylbenzhydrazide
PubChem CID 82110
IUPAC Name 2-methylbenzohydrazide
SMILES CC1=CC=CC=C1C(=O)NN

Specifications

Melting Point 122°C to 124°C
Quantity 5 g
Beilstein 1862809
Formula Weight 150.18
Percent Purity ≥98%
Chemical Name or Material o-Toluic hydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.