missing translation for 'onlineSavingsMsg'
Learn More

o-Toluidine, 99%, Thermo Scientific Chemicals

Product Code. 11400154
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400154 1000 g 1000g
11410154 250 g 250g
11420154 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11400154 Supplier Thermo Scientific Alfa Aesar Supplier No. A13602.0B
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

o-Toluidine is used in the production of dyes, as a presumptive test for blood in forensic science. And it is also used in determination of glucose in biological materials, as a precursor in the synthesis of poly(o-toluidine) and copper nanoparticle composite material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Toluidine is used in the production of dyes, as a presumptive test for blood in forensic science. And it is also used in determination of glucose in biological materials, as a precursor in the synthesis of poly(o-toluidine) and copper nanoparticle composite material.

Solubility
Soluble in water(15g/L).

Notes
Air & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-53-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00007730
InChI Key RNVCVTLRINQCPJ-UHFFFAOYSA-N
Synonym o-toluidine, 2-toluidine, o-tolylamine, 2-aminotoluene, 2-methylbenzenamine, ortho-toluidine, o-methylaniline, benzenamine, 2-methyl, o-aminotoluene, o-methylbenzenamine
PubChem CID 7242
ChEBI CHEBI:66892
IUPAC Name 2-methylaniline
SMILES CC1=CC=CC=C1N

Specifications

Melting Point -24°C
Density 1.004
Boiling Point 199°C to 200°C
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.572
Quantity 1000 g
UN Number UN1708
Beilstein 741981
Sensitivity Air and light sensitive
Merck Index 14,9536
Solubility Information Soluble in water(15g/L).
Formula Weight 107.16
Percent Purity 99%
Chemical Name or Material o-Toluidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.