missing translation for 'onlineSavingsMsg'
Learn More

o-Tolyl benzoate, 99%, Thermo Scientific™

Product Code. 11372647
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372647 100 g 100g
11362647 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372647 Supplier Thermo Scientific Alfa Aesar Supplier No. L11834.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-02-7
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00041330
InChI Key YPMKPCAILYDVBN-UHFFFAOYSA-N
Synonym o-tolyl benzoate, benzoic acid, o-tolyl ester, o-tolylbenzoate, benzoic acid, 2-methylphenyl ester, toluene benzoate, acmc-1az7o, 2-methylphenyl benzoate, benzoic acid o-tolyl ester, ortho-tolyl benzoate
PubChem CID 69231
IUPAC Name (2-methylphenyl) benzoate
SMILES CC1=CC=CC=C1OC(=O)C1=CC=CC=C1

Specifications

Density 1.091
Boiling Point 170°C to 172°C (11 mmHg)
Refractive Index 1.57
Quantity 100 g
Beilstein 1873222
Formula Weight 212.25
Percent Purity 99%
Chemical Name or Material o-Tolyl benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.