missing translation for 'onlineSavingsMsg'
Learn More

o-Tolyl isothiocyanate, 99%

Product Code. 10177833 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
10177833 5 g 5g
10289583 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 10177833

Brand: Thermo Scientific Acros 292650050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 614-69-7 | C8H7NS | 149.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-69-7
MDL Number MFCD00004802
InChI Key JYKYYPPZLPVIBY-UHFFFAOYSA-N
Synonym 2-methylphenyl isothiocyanate, o-tolyl isothiocyanate, 2-tolyl isothiocyanate, o-tolylisothiocyanate, benzene, 1-isothiocyanato-2-methyl, 1-isothiocyanato-2-methyl-benzene, 2-methylphenylisothiocyanate, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, 2-methylbenzenisothiocyanate
PubChem CID 69195
IUPAC Name 1-isothiocyanato-2-methylbenzene
SMILES CC1=CC=CC=C1N=C=S

Specifications

Color Yellow
Density 1.1100g/mL
Boiling Point 239°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H7NS
Linear Formula CH3C6H4NCS
Refractive Index 1.6351 to 1.6371
Quantity 5 g
Specific Gravity 1.11
Solubility Information Solubility in water: may decompose
Molecular Weight (g/mol) 149.22
Formula Weight 149.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material o-Tolyl isothiocyanate
Show More Show Less
compliance-icons
Product Identifier
  • o-Tolyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.