missing translation for 'onlineSavingsMsg'
Learn More

o-Xylylene dibromide, 97%

Product Code. 11423467
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423467 5 g 5g
11433467 25 g 25g
11443467 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423467 Supplier Thermo Scientific Alfa Aesar Supplier No. A17241.06
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 91-13-4 | C8H8Br2 | 263.96 g/mol

1,2-Bis(bromomethyl)benzene is used in the synthesis of isothioureas involved in the inhibition of human nitric oxide synthases. Also used in the synthesis of tridentate carbene ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(bromomethyl)benzene is used in the synthesis of isothioureas involved in the inhibition of human nitric oxide synthases. Also used in the synthesis of tridentate carbene ligands.

Solubility
Solubility in toluene is almost transparent and ethanol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-13-4
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000175
InChI Key KGKAYWMGPDWLQZ-UHFFFAOYSA-N
Synonym 1,2-bis bromomethyl benzene, o-xylylene dibromide, alpha,alpha'-dibromo-o-xylene, benzene, 1,2-bis bromomethyl, o-bis bromomethyl benzene, o-xylylene bromide, 1,2-dibenzyl bromide, alpha,alpha'-dibromo-o-xylol, a,a'-dibromo-o-xylene, ccris 1776
PubChem CID 66665
IUPAC Name 1,2-bis(bromomethyl)benzene
SMILES C1=CC=C(C(=C1)CBr)CBr

Specifications

Melting Point 92°C to 96°C
Density 1.96
Quantity 5 g
UN Number UN3261
Beilstein 637159
Formula Weight 263.96
Percent Purity 97%
Chemical Name or Material o-Xylylene dibromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.