missing translation for 'onlineSavingsMsg'
Learn More

pH Buffer Pouches, Oakton™

Product Code. 17977242
Change view
Click to view available options
Quantity:
20 Ea.
pH:
4.01
10.00
Unit Size:
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity pH Unit Size
17977242 20 Ea. 10.00 Pack of 20
17967242 20 Ea. 4.01 Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17977242 Supplier Oakton Supplier No. 3565303
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oakton™ pH buffer pouches; package of 20 Economical pouches are accurate and convenient.±0.01 accuracy at 77°°F (25°°C)

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 10.00
Physical Form Liquid
Quantity 20 Ea.
Solution Type pH Buffer
Grade Not Available
Chemical Name or Material Oakton pH buffer pouches
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.