missing translation for 'onlineSavingsMsg'
Learn More

buffer pouches, Cole-Parmer Instrument Company, Oakton™

Product Code. 17907242
Change view
Click to view available options
Quantity:
20 Pouches
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17907242 20 Pouches Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17907242 Supplier Oakton Supplier No. 3565302
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Economical pouches are accurate and convenient.

Applications: Electrochemistry, Laboratory Research, General Research, Pharmaceutical

Specifications

CAS 7778-77-0
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 7
Physical Form Liquid
Quantity 20 Pouches
Solution Type pH Buffer
Molecular Formula H2KO4P
MDL Number MFCD00011401,MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES [K+].OP(O)([O-])=O
IUPAC Name potassium dihydrogen phosphate
Molecular Weight (g/mol) 136.08
PubChem CID 516951
ChEBI CHEBI:63036
Chemical Name or Material OAKTONpH buffer pouches
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.