missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OASIS/CREB3L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
292.00 € - 572.00 €
Specifications
| Antigen | OASIS/CREB3L1 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18425131
|
Novus Biologicals
NBP1-82503-25ul |
25 μL |
292.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18290246
|
Novus Biologicals
NBP1-82503 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OASIS/CREB3L1 Polyclonal specifically detects OASIS/CREB3L1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| OASIS/CREB3L1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 90993 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LNESDFLNNAHFPEHLDHFTENMEDFSNDLFSSFFDDPVLDEKSPLLDMELDSPTPGIQAEHSYSLSGDSAPQSPLVPIKMEDTTQDAEHGAWALGHKLCSIMVKQE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| BBF-2 homolog, cAMP responsive element binding protein 3-like 1, cAMP-responsive element-binding protein 3-like protein 1, OASIScyclic AMP-responsive element-binding protein 3-like protein 1, old astrocyte specifically induced substance, Old astrocyte specifically-induced substance | |
| CREB3L1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title