Learn More
OATP1B3/SLCO1B3/OATP8 Antibody (CL3771) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61637CL1
Product Code. 30289552
  • 1175.00 € / 0.10mL
Description
OATP1B3/SLCO1B3/OATP8 Monoclonal antibody specifically detects OATP1B3/SLCO1B3/OATP8 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OATP1B3/SLCO1B3/OATP8
Monoclonal
CoraFluor 1
Liver-specific organic anion transporter 2, liver-specific organic anion transporter 3TM13, LST2, LST-2, LST3, OATP1B3LST-3TM13, OATP8, OATP-8, Organic anion transporter 8, organic anion transporter LST-3c, Organic anion-transporting polypeptide 8, SLC21A8, solute carrier family 21 (organic anion transporter), member 8, Solute carrier family 21 member 8, solute carrier organic anion transporter family member 1B3, solute carrier organic anion transporter family, member 1B3
This antibody was developed using a recombinant protein derived from Q9NPD5, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
28234
Store at 4⪚C in the dark. Do not freeze.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL3771
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified