missing translation for 'onlineSavingsMsg'
Learn More

Ochratoxin A, 99+%

Product Code. 10470691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mg
Packaging:
Glass bottle
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10470691 5 mg Glass bottle 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10470691 Supplier Thermo Scientific Acros Supplier No. 227400050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ochratoxin A, CAS # 303-47-9, is a mycotoxin produced by certain fungi. | CAS: 303-47-9 | C20H18ClNO6 | 403.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Ochratoxin A is a mycotoxin of fungal origin, particularly from Aspergillus and Penicillium species, commonly found in spoiled batches of grains, beans, and nuts
• Ochratoxin A is a known mutagen and potential carcinogen; suspected to affect fetal development and promote the development of urinary tract tumors
Applications
• Ochratoxin A can be used as a calcium channel blocker in cells
• May be useful for developmental research studies
• Intended for research applications only

TRUSTED_SUSTAINABILITY

Specifications

CAS 303-47-9
CAS Min % 99.0
CAS Max % 100.0
Melting Point 169.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Molecular Formula C20H18ClNO6
MDL Number MFCD00078079
Quantity 5 mg
Beilstein 18, V,9, 56
Merck Index 15, 6829
Synonym ochratoxin a, antibiotic 9663, phenylalanine-ochratoxin a, 3r14s-ochratoxin a, ochratoxin a-bsa conjugate from aspergillus ochraceus, --n-5-chloro-8-hydroxy-3-methyl-1-oxo-7-isochromanyl carbonyl-3-phenylalanine, n-3r-5-chloro-8-hydroxy-3-methyl-1-oxo-7-isochromanyl carbonyl-3-phenyl-l-alanine, unii-1779sx6luy, l-phenylalanine, n-5-chloro-3,4-dihydro-8-hydroxy-3-methyl-1-oxo-1h-2-benzopyran-7-yl carbonyl-, r, ccris 2382
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in chloroform, diethyl ether, benzene,, ethanol, methanol
InChI Key RWQKHEORZBHNRI-BMIGLBTASA-N
SMILES CC1CC2=C(C=C(C(=C2C(=O)O1)O)C(=O)NC(CC3=CC=CC=C3)C(=O)O)Cl
IUPAC Name (2S)-2-[[(3R)-5-chloro-8-hydroxy-3-methyl-1-oxo-3,4-dihydroisochromene-7-carbonyl]amino]-3-phenylpropanoic acid
Molecular Weight (g/mol) 403.82
PubChem CID 442530
ChEBI CHEBI:7719
Formula Weight 403.82
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Ochratoxin A, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ochratoxin A
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.