missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCRL Antibody (S166A-26), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42193-0.025mg
This item is not returnable.
View return policy
Description
OCRL Monoclonal antibody specifically detects OCRL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| OCRL | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence | |
| Q01968 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| S166A-26 | |
| Unconjugated | |
| PBS (pH 7.4), 50% Glycerol | |
| EC 3.1.3.36, INPP5F, LOCR, Lowe oculocerebrorenal syndrome protein, NPHL2, OCRL1inositol polyphosphate 5-phosphatase OCRL-1, oculocerebrorenal syndrome of Lowe, phosphatidylinositol polyphosphate 5-phosphatase | |
| Fusion protein amino acids 1-901 (full-length) of human INPP5F. Rat: 93% identity (845/904 amino acids identical). Mouse: 91% identity (824/901 amino acids identical) ∽50% identity with INPP5b. | |
| 0.025 mg | |
| Golgi Apparatus Markers | |
| 4952 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction