Learn More
OCT4 Mouse anti-Human, Mouse, Rat, Clone: OTI7H1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF506798
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Oct1 or POU2F1 (Pou domain, class 2, transcription factor 1) is a transcription factor that binds to the octamer motif (5′-ATTTGCAT-3′) and activates the promoters of the genes for some small nuclear RNAs (snRNA) and of genes such as those for histone H2B and immunoglobulins. It modulates transcription transactivation by NR3C1, AR, and PGR and interacts with NR3C1, AR, and PGR. Oct1 is ubiquitous and belongs to the POU transcription factor family class 2 subfamily. Oct1 contains 1 homeobox domain.Specifications
| OCT4 | |
| Monoclonal | |
| Unconjugated | |
| POU5F1 | |
| OCT3, OCT4, OTF-3, OTF3, OTF4, Oct-3, Oct-4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 18999, 294562, 5460 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI7H1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P20263, Q01860 | |
| POU5F1 | |
| Full length human recombit protein of human POU5F1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.