missing translation for 'onlineSavingsMsg'
Learn More

Octafluoronaphthalene, 96%

Product Code. 11490287
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490287 1 g 1g
11400297 5 g 5g
11438710 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490287 Supplier Thermo Scientific Alfa Aesar Supplier No. B21699.03
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 313-72-4 | C10F8 | 272.097 g/mol

It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water. Solubility in toluene, almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313-72-4
Molecular Formula C10F8
Molecular Weight (g/mol) 272.097
MDL Number MFCD00014307
InChI Key JDCMOHAFGDQQJX-UHFFFAOYSA-N
Synonym octafluoronaphthalene, perfluoronaphthalene, naphthalene, octafluoro, octafluoronapthalene, octafiuoronaphthalene, acmc-1cr7y, octafluoronaphthalene 1g, octafluoronaphthalene radical anion
PubChem CID 67564
IUPAC Name 1,2,3,4,5,6,7,8-octafluoronaphthalene
SMILES C12=C(C(=C(C(=C1F)F)F)F)C(=C(C(=C2F)F)F)F

Specifications

Melting Point 84°C to 89°C
Boiling Point 209°C
Quantity 1 g
Beilstein 1915630
Solubility Information Insoluble in water. Solubility in toluene,almost transparent.
Formula Weight 272.1
Percent Purity 96%
Chemical Name or Material Octafluoronaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.