missing translation for 'onlineSavingsMsg'
Learn More

Octyltriphenylphosphonium Bromide, TRC

Product Code. 30426487
Change view
Click to view available options
Quantity:
2.5 g
10 g
25 g
Unit Size:
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426487 10 g -
30413697 2.5 g 2.5g
30408959 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426487 Supplier Toronto Research Chemicals Supplier No. TRCO24139510G
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42036-78-2
Molecular Formula C26H32BrP
Molecular Weight (g/mol) 455.41
IUPAC Name octyl(triphenyl)phosphanium;bromide
SMILES [Br-].CCCCCCCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

Specifications

Quantity 10 g
InChI Formula InChI=1S/C26H32P.BrH/c1-2-3-4-5-6-16-23-27(24-17-10-7-11-18-24,25-19-12-8-13-20-25)26-21-14-9-15-22-26;/h7-15,17-22H,2-6,16,23H2,1H3;1H/q+1;/p-1
Formula Weight 454.1425
Chemical Name or Material Octyltriphenylphosphonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.