missing translation for 'onlineSavingsMsg'
Learn More

Ohio Pesticide Mixture 335 10-100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30323302
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323302 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323302 Supplier Dr. Ehrenstorfer Supplier No. DREA50000335AL
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Abamectin∣Aldicarb∣Bifenazate∣Cyfluthrin∣Daminozide∣Diazinon∣Dichlorvos∣Dimethoate∣Etoxazole∣Flonicamid∣Fludioxonil∣Imidacloprid∣Myclobutanil∣Paclobutrazol∣Piperonyl butoxide∣Pyrethrins∣Spinosad∣Spirotetramat∣Thiamethoxam∣Trifloxystrobin
CAS 71751-41-2, 116-06-3, 149877-41-8, 68359-37-5, 1596-84-5, 333-41-5
Analyte or Component Names Abamectin∣Aldicarb∣Bifenazate∣Cyfluthrin∣Daminozide∣Diazinon∣Dichlorvos∣Dimethoate∣Etoxazole∣Flonicamid∣Fludioxonil∣Imidacloprid∣Myclobutanil∣Paclobutrazol∣Piperonyl butoxide∣Pyrethrins∣Spinosad∣Spirotetramat∣Thiamethoxam∣Trifloxystrobin
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Abamectin 10 μg/mL∣Aldicarb 10 μg/mL∣Cyfluthrin 10 μg/mL∣Daminozide 10 μg/mL∣Dichlorvos 10 μg/mL∣Dimethoate 10 μg/mL∣Etoxazole 10 μg/mL∣Fludioxonil 10 μg/mL∣Imidacloprid 10 μg/mL∣Myclobutanil 10 μg/mL∣Paclobutrazol 10 μg/mL∣Spinosad 10 μg/mL∣Bifenazate 20 μg/mL∣Thiamethoxam 20 μg/mL∣Trifloxystrobin 20 μg/mL∣Flonicamid 30 μg/mL∣Pyrethrins 50 μg/mL∣Diazinon 100 μg/mL∣Piperonyl butoxide 100 μg/mL∣Spirotetramat 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.