missing translation for 'onlineSavingsMsg'
Learn More

Ohio Residual Solvent Mixture 1000 μg/mL in Triacetin, Dr. Ehrenstorfer

Product Code. 30328653
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328653 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328653 Supplier Dr. Ehrenstorfer Supplier No. DRES50000004TN

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

CAS 67-64-1, 71-43-2, 64-17-5, 67-63-0, 106-97-8, 142-82-5
Quantity 5 x 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Triacetin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.