Learn More
Fisher Chemical Oil Test Strips
Product Code. 31937638 Shop All Fisher Chemical Products
Change view
Click to view available options
Detects:
Oil
Unit Size:
Pack of 100
Product Code. Detects Unit Size
31937638 Oil Pack of 100
1 options
Product Code. 31937638 Supplier Fisher Chemical Supplier No. PF0546
  • ONLINE EXCLUSIVE PRICE
    42.38 €
    32.73 € / Pack of 100
    Save 9.65 €
    22% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Oil Test Strips provide precise readings within seconds, helping kitchens monitor oil degradation, extend oil life, and maintain compliance with food safety regulations.
  • Fast and easy to use - No training or special equipment required
  • Supports HACCP and food safety compliance
  • Improves food presentation and taste
  • Reduces oil waste and fryer maintenance costs
  • Trusted by chefs and kitchen managers across the UK and Europe
  • Easy-to-Use: Designed with simplicity in mind, these test strips offer a hassle-free testing experience. With a user-friendly dip-and-read method, chefs and home cooks can quickly assess the quality of their cooking oil without the need for specialized equipment or extensive training.
  • High Sensitivity: Engineered to detect various indicators of cooking oil degradation, these test strips offer unparalleled sensitivity. From Free Fatty Acid (FFA) levels to Total Polar Compounds (TPC), users can obtain comprehensive insights into the quality and freshness of their cooking oil.
  • Rapid Results: With rapid response times, users can obtain test results within 30 seconds, allowing for timely intervention and proactive oil management. Whether it's determining the optimal time for oil replacement or monitoring oil stability during frying, these test strips empower users to maintain peak oil performance.
  • Versatility: Suitable for a wide range of cooking oils, including vegetable oil, olive oil, canola oil, and more, these testing strips offer versatility and adaptability to diverse culinary applications.
  • Cost-Effective: By enabling precise oil management and extending the lifespan of cooking oil, these test strips help reduce waste and optimize operational costs for food service establishments. Their affordability and efficiency make them a valuable investment for kitchens of all sizes.
Specifications
Detects Oil
Manufacturing Origin United Kingdom
Product Type Oil Test Strip
Test Type Qualitative QC
Quantity 1 Ea.