missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olaparib, MedChemExpress
Description
Olaparib (AZD2281
Specifications
Specifications
| Chemical Name or Material | Olaparib |
| CAS | 763113-22-0 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H23FN4O3 |
| Quantity | 10 mg |
| Synonym | AZD2281 KU0059436 |
| Solubility Information | DMSO : 100 mg/mL (230.17 mM; Need ultrasonic) ∣DMF : 50 mg/mL (115.09 mM; Need ultrasonic) ∣Ethanol : 3.12 mg/mL (7.18 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C(C1=CC(CC(C2=C3C=CC=C2)=NNC3=O)=CC=C1F)N4CCN(C(C5CC5)=O)CC4 |
| Molecular Weight (g/mol) | 434.46 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?