missing translation for 'onlineSavingsMsg'
Learn More

Oleacein, 90%, TRC

Product Code. 17998964
Change view
Click to view available options
Quantity:
1 mg
10 mg
100 mg
Unit Size:
100mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17998964 10 mg 10mg
17968774 100 mg 100mg
17998204 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17998964 Supplier Toronto Research Chemicals Supplier No. TRCO52455010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149183-75-5
Molecular Formula C17H20O6
Molecular Weight (g/mol) 320.34
Synonym (3S,4E)-4-Formyl-3-(2-oxoethyl)-4-hexenoic Acid 2-(3,4-Dihydroxyphenyl)ethyl Ester, [S-(E)]-4-Formyl-3-(2-oxoethyl)-4-hexenoic Acid 2-(3,4-Dihydroxyphenyl)ethyl Ester
IUPAC Name 2-(3,4-dihydroxyphenyl)ethyl (E,3S)-4-formyl-3-(2-oxoethyl)hex-4-enoate
SMILES C\C=C(\C=O)/[C@@H](CC=O)CC(=O)OCCc1ccc(O)c(O)c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H20O6/c1-2-13(11-19)14(5-7-18)10-17(22)23-8-6-12-3-4-15(20)16(21)9-12/h2-4,7,9,11,14,20-21H,5-6,8,10H2,1H3/b13-2-/t14-/m0/s1
Formula Weight 320.126
Chemical Name or Material Oleacein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.