missing translation for 'onlineSavingsMsg'
Learn More

Oligomycin

Product Code. 15414389
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414389 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15414389 Supplier Thermo Scientific Alfa Aesar Supplier No. J61898.MA
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oligomycin, CAS # 1404-19-9, is a macrolide antibiotic that inhibits oxidative phosphorylation through blocking mitochondrial ATP synthase. | CAS: 1404-19-9 | C45H74O11 | 791.08 g/mol

Inhibits respiration in mitochondria. Also inhibits ATP synthase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Oligomycin is a macrolide produced by Streptomyces diastatochromogenes that is toxic to other organisms
  • Oligomycin blocks oxidative phosphorylation through inhibition of ATP synthase. The compound binds to the proton channel of ATP synthase and blocks mitochondrial proton import, resulting in accumulation of cytoplasmic protons
  • Oligomycin also inhibits Na+/K+ exchange through Na+ occlusion within the Na/K-ATPase
  • Oligomycin is provided as a mixture of A, B, and C isomers

Application

  • Oligomycin inhibits respiration in mitochondria. It also inhibits ATP synthase
  • Oligomycin can induce apoptosis in cultured human hepatocarcinoma and other mammalian cells in laboratory settings
TRUSTED_SUSTAINABILITY

Specifications

Type Oligomycin
Physical Form Powder
CAS 1404-19-9
Color White
Quantity 10 mg
Molecular Formula C45H74O11
MDL Number MFCD00065705,MFCD01779388
Beilstein 7615308
Merck Index 14,6833
Solubility Information Sparingly soluble in water; Soluble in DMSO (20mg/ml) and ethanol (30mg/ml) /n
InChI Key MNULEGDCPYONBU-ZUSSGZTJNA-N
SMILES CCC1CCC2OC3(CCC(C)C(CC(C)O)O3)C(C)C(OC(=O)\C=C\C(C)C(O)C(C)C(=O)C(C)C(O)C(C)C(=O)C(C)(O)C(O)C(C)C\C=C\C=C/1)C2C
IUPAC Name (4E,18E,20Z)-22-ethyl-7,11,14,15-tetrahydroxy-6'-(2-hydroxypropyl)-5',6,8,10,12,14,16,28,29-nonamethyl-2,26-dioxaspiro[bicyclo[23.3.1]nonacosane-27,2'-oxane]-4,18,20-triene-3,9,13-trione
Molecular Weight (g/mol) 791.08
PubChem CID 78358496
Formula Weight 791.07
Chemical Name or Material Oligomycin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.