missing translation for 'onlineSavingsMsg'
Learn More

Olopatadine hydrochloride, MedChemExpress

Product Code. 16484357
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484357 50 mg 50mg
16418488 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16484357 Supplier Medchem Express Supplier No. HYB0426A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Olopatadine hydrochloride (ALO4943A) is a histamine blocker used to treat allergic conjunctivitis.

Olopatadine hydrochloride (ALO4943A) is a histamine blocker used to treat allergic conjunctivitis.

Specifications

Chemical Name or Material Olopatadine hydrochloride
CAS 140462-76-6
Color White
Purity Grade Notes Research
Molecular Formula C21H24ClNO3
Quantity 50 mg
Synonym ALO4943A KW4679
Solubility Information DMSO : 50 mg/mL (133.74 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (17.84 mM; Need ultrasonic)
SMILES O=C(O)CC1=CC=C(C/2=C1)OCC3=CC=CC=C3C2=C\CCN(C)C.Cl
Molecular Weight (g/mol) 373.87
Formula Weight 373.87
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.