missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OMA1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94293-0.1ml
This item is not returnable.
View return policy
Description
OMA1 Polyclonal antibody specifically detects OMA1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| OMA1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 2010001O09Rik, DAB1, EC 3.4.24.-, metalloprotease related protein 1, Metalloprotease-related protein 1, MPRP1, MPRP-1mitochondrial, OMA1 homolog, zinc metallopeptidase (S. cerevisiae), overlapping activity with M-AAA protease, YKR087C, ZMPOMA1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 360-524 of human OMA1 (NP_660286.1). AICPRDSLALLCQWIQSKLQEYMFNRPYSRKLEAEADKIGLLLAAKACADIRASSVFWQQMEFVDSLHGQPKMPEWLSTHPSHGNRVEYLDRLIPQALKIREMCNCPPLSNPDPRLLFKLSTKHFLEESEKEDLNITKKQKMDTLPIQKQEQIPLTYIVEKRTGS | |
| 0.1 mL | |
| Cell Biology, Endocrinology | |
| 115209 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction