Learn More
Invitrogen™ Omburtamab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559613
 View more versions of this product
Product Code. 30283254
  • 477.00 € / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Omburtamab is a monoclonal antibody that targets the B7-H3 antigen expressed in a variety of human solid tumors. Omburtamab (alias: 8H9, former name: Burtomab) is a monoclonal antibody that is linked to a radioisotope and can bind to the B7-H3 antigen on the surface of neuroblastoma cells. It is injected directly into the cerebrospinal fluid to bypass the blood-brain barrier and accurately target neuroblastoma in the central nervous system to avoid damage to other tissues (such as learning disabilities and developmental delays often caused by conventional radiotherapy outside the brain).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Omburtamab
Recombinant Monoclonal
Unconjugated
8H9; Mab 8H9
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized