missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OMP Recombinant Rabbit Monoclonal Antibody (K01_1W09)
Descripción
OMP Recombinant Monoclonal Antibody for Western Blot
Olfactory marker protein (OMP) is an abundant, 19-kDa, cytosolic protein that is almost exclusively expressed in mature, functioning, olfactory neurons but not in the neural precursor basal cells. The OMP gene structure and protein sequence are highly conserved between mouse, rat and human. Its tissue-specific expression in the receptor cells together with the results of the mouse knockout studies show that OMP represents a novel modulatory component of the odor detection/signal transduction cascade. May act as a modulator of the olfactory signal-transduction cascade. Interacts with BEX1 and BEX2. Uniquely associated with mature olfactory receptor neurons. Belongs to the olfactory marker protein family.
Especificaciones
Especificaciones
| Antigen | OMP |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1W09 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | Omp |
| Gene Accession No. | P08523, Q64288 |
| Gene Alias | olfactory marker protein; Olfactory neuronal-specific protein; OMP |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?