missing translation for 'onlineSavingsMsg'
Learn More

ON-013100, MedChemExpress

Product Code. 16485536
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485536 100 mg 100mg
16437427 5 mg 5mg
16421189 1 mg 1mg
16411007 50 mg 50mg
16404718 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16485536 Supplier Medchem Express Supplier No. HY112822
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress ON-013100, an antineoplastic drug, acts a mitotic inhibitor that could inhibit Cyclin D1 expression.

ON-013100, an antineoplastic drug, acts a mitotic inhibitor that could inhibit Cyclin D1 expression.

Specifications

Chemical Name or Material ON-013100
CAS 865783-95-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H22O7S
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (63.38 mM; Need ultrasonic)
SMILES OC1=CC(CS(=O)(/C=C/C2=C(OC)C=C(OC)C=C2OC)=O)=CC=C1OC
Molecular Weight (g/mol) 394.44
Formula Weight 394.44
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.