missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oncomodulin/OCM2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 470.00 €
Specifications
| Antigen | Oncomodulin/OCM2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602092
|
Novus Biologicals
NBP2-95179-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18662182
|
Novus Biologicals
NBP2-95179-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Oncomodulin/OCM2 Polyclonal antibody specifically detects Oncomodulin/OCM2 in Rat samples. It is validated for Western BlotSpecifications
| Oncomodulin/OCM2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat | |
| OCM2, oncomodulin 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-109 of human OCM2 (NP_006179.2). MSITDVLSADDIAAALQECQDPDTFEPQKFFQTSGLSKMSASQVKDVFRFIDNDQSGYLDEEELKFFLQKFESGARELTESETKSLMAAADNDGDGKIGAEEFQEMVHS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 4951 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title