missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ONECUT2/OC-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
320.00 € - 624.00 €
Specifications
| Antigen | ONECUT2/OC-2 |
|---|---|
| Applications | Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18406791
|
Novus Biologicals
NBP2-31658-25ul |
25 μL |
320.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18123265
|
Novus Biologicals
NBP2-31658 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ONECUT2/OC-2 Polyclonal specifically detects ONECUT2/OC-2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ONECUT2/OC-2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| O95948 | |
| 9480 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MLTRGEQHLSRGLGTPPAAMMSHLNGLHHPGHTQSHGPVLAPSRERPPSSSSGSQVATSGQLEE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| one cut domain family member 2, OC2, OC-2, one cut homeobox 2 | |
| ONECUT2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title