missing translation for 'onlineSavingsMsg'
Learn More

ONO-4059 analog, MedChemExpress

Product Code. 16480118
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480118 5 mg 5mg
16473616 25 mg 25mg
16467556 100 mg 100mg
16433667 50 mg 50mg
16417658 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16480118 Supplier Medchem Express Supplier No. HY18951

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ONO-4059 analog is the analog of ONO-4059, ONO-4059 is a highly potent and selective Btk inhibitor.

ONO-4059 analog is the analog of ONO-4059, ONO-4059 is a highly potent and selective Btk inhibitor.

Specifications

Chemical Name or Material ONO-4059 analog
CAS 1351635-67-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H24N6O3
Quantity 5 mg
Solubility Information DMSO : ≥ 29 mg/mL (63.53 mM)
SMILES O=C1N([C@H]2CCCN(C(C=C)=O)C2)C3=NC=NC(N)=C3N1C4=CC=C(OC5=CC=CC=C5)C=C4
Molecular Weight (g/mol) 456.5
Formula Weight 456.5
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.