missing translation for 'onlineSavingsMsg'
Learn More

ONO-7300243, MedChemExpress

Product Code. 16475937
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16475937 5 mg 5mg
16463208 10 mg 10mg
16459756 50 mg 50mg
16442676 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16475937 Supplier Medchem Express Supplier No. HY100882
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ONO-7300243 is a novel, potent lysophosphatidic acid receptor 1 (LPA1) antagonist with IC50 of 0.16 μM.

ONO-7300243 is a novel, potent lysophosphatidic acid receptor 1 (LPA1) antagonist with IC50 of 0.16 μM.

Specifications

Chemical Name or Material ONO-7300243
CAS 638132-34-0
Color White
Purity Grade Notes Research
Molecular Formula C28H31NO5
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (216.66 mM; Need ultrasonic)
SMILES O=C(O)CC1=CC=C(CN(C(C2=CC(OC)=C(C)C(OC)=C2)=O)CCCC3=CC=CC=C3)C=C1
Molecular Weight (g/mol) 461.55
Formula Weight 461.55
Percent Purity 98.14%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.