missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sifalimumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558636
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Sifalimumab is a fully human monoclonal antibody targeting interferon-alpha. The levels of interferon-alpha are elevated in many patients with active systemic lupus erythematosus (SLE, or lupus) and other autoimmune disorders, and may be associated with disease activity. Sifalimumab may suppress the abnormal immune activity associated with lupus by binding to multiple interferon-alpha subtypes seen in the serum of lupus patients.
Specifications
| Sifalimumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MDX-1103; MEDI-545 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction