missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Op-Air™ Anti-fog Medical Mask with Foam Band
GREENER_CHOICE

Product Code. 17700189 Shop All Medicom Products
Change view
Click to view available options
Op-Air Anti-fog Medical Mask with Foam Band; Certifications/Compliance (Europe): (EU) 2017/745, EN 14683, ISO 13485, ISO 9001; Color: Blue; Disposable: Yes; Fastening Type: Ties; Style: Pleated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17700189 Op-Air Anti-fog Medical Mask with Foam Band; Certifications/Compliance (Europe): (EU) 2017/745, EN 14683, ISO 13485, ISO 9001; Color: Blue; Disposable: Yes; Fastening Type: Ties; Style: Pleated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 17700189 Supplier Kolmi Hopen™ Supplier No. M21101M30
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

The unique addition of the foam band to this mask provides extra comfort to the wearer on top of fog prevention.

  • High filtration media (polypropylene)
  • Layers (interior to exterior): thermobonded polypropylene, melt blown polypropylene, spunbonded polypropylene
  • Soft, non-woven ties and edges
  • Anatomical folding limits exposure to face
  • Integrated anti-fog band and nose piece
  • Fiberglass-free filter
  • Produced in ISO 8 clean room (particle and microbiological control)

Specifications

Color Blue
Fastening Type Ties
Material Polypropylene
Style Pleated
Certifications/Compliance (Europe) (EU) 2017/745, EN 14683, ISO 13485, ISO 9001
Disposable Single-use
For Use With (Application) Medical
Packaging Quantity 50 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.