missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Op-Air™ One Anti-glare Medical Mask

Product Code. 17750189 Shop All Medicom Products
Change view
Click to view available options
Op-Air One Anti-glare Medical Mask; Certifications/Compliance (Europe): (EU) 2017/745, EN 14683, ISO 13485, ISO 9001; Color: Blue; Disposable: Yes; Fastening Type: Ties; Material: Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17750189 Op-Air One Anti-glare Medical Mask; Certifications/Compliance (Europe): (EU) 2017/745, EN 14683, ISO 13485, ISO 9001; Color: Blue; Disposable: Yes; Fastening Type: Ties; Material: Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 17750189 Supplier Kolmi Hopen™ Supplier No. M24101N30
  • ONLINE EXCLUSIVE PRICE
    8.60 €
    7.64 € / Pack of 50
    Save 0.96 €
    11% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Be prepared for long hours of wear with this anti-glare medical mask complete with anti-reflective tape.

  • High filtration media (polypropylene)
  • Layers (interior to exterior): wet laid cellulose, melt blown polypropylene, spunbonded polypropylene
  • Soft, non-woven mask ties and edges
  • Anatomical folding limits exposure to face
  • Integrated nose bar and anti-reflective tape to limit glare phenomena in operating room
  • Fiberglass-free filter
  • Produced in ISO 8 clean room (particle and microbiological control)

Specifications

Color Blue
Fastening Type Ties
Material Polypropylene
Style Pleated
Certifications/Compliance (Europe) (EU) 2017/745, EN 14683, ISO 13485, ISO 9001
Disposable Single-use
For Use With (Application) Medical
Packaging Quantity 50 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.