Learn More
Kolmi™ Op Air Pro Oxygen FFP2 NR D Mass Balance Masks
Product Code. 17774789 Shop All Medicom Products
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17774789 Medium Pack of 50
17794789 Small Pack of 50
17784789 Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17774789 Supplier Kolmi™ Supplier No. M52014SWHMB
  • ONLINE EXCLUSIVE PRICE
    22.37 €
    19.35 € / Pack of 50
    Save 3.02 €
    13% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Op Air Pro Oxygen FFP2 masks are easy to put on with high breathability due to high quality raw material. Made with the Mass Balance approach from recycled materials.
  • Filtering half mask, efficiency class FFP2 NR D developed to protect against solid and liquid aerosols and particles
  • Type IIR certified, it protects the environment against droplets emitted by the wearer and it protects from potential body fluid splashes
  • Knitted headloops ultrasonically welded for optimal comfort
  • Fits perfectly to face to limit inward leakage thanks to its shape and soft materials
  • Hypoallergenic and has been dermatologically tested
  • Packaged in individual polybags
  • Made in France
Specifications
Certifications/Compliance EN 149: 2001 + A1: 2009, DM Class I, EN 14683: 2019 + AC: 2019
Color White
Size Medium
Style Pouch
Strap Attachment Type Headband