Learn More
Kolmi Hopen™ Op-R™ Type II Medical Masks with Earloops
Pleated medical mask with anti-fog nose clip for patients and healthcare workers.
Brand:  Kolmi Hopen™ M13201-30
Additional Details : Weight : 3.70500kg
 View more versions of this product
Product Code. 17740179
  • 5.20 € / Pack of 60
Estimated Shipment: 19-07-2024
to see stock.
Add to basket
Description
Description
  • High filtration media (polypropylene)
  • Two wide and comfortable elastic bands
  • Non-woven, soft bias binding
  • Integrated anti-fog nose clip
  • Filter without fiberglass
  • Produced in ISO 8 clean room (particulate and microbiological control)
Specifications
Specifications
Green
Polypropylene
CE, ISO 13485, (EU) 2017/745, EN 14683, EN ISO 14971
Medical
Earloops
Pleated
Yes
60 Pk.