missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPA1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-55290AF750
This item is not returnable.
View return policy
Description
OPA1 Polyclonal specifically detects OPA1 in Human, Mouse, Rat, Porcine, Chicken, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting.
Specifications
| OPA1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting | |
| FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, MGM1, mitochondrial dynamin-like GTPase, NPGlargeG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1 | |
| Rabbit | |
| Affinity Purified | |
| Apoptosis, Mitochondrial Fusion Proteins, Mitochondrial Mediated Pathway, Neurodegeneration, Neuroscience, Sensory Systems, Vision | |
| 4976.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoblot | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| OPA1 | |
| A synthetic peptide made to an internal region within residues 500-600 of human OPA1. [Swiss-Prot: O60313] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Pig, Chicken, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction